Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS10840EA 4940 CZK
AGRIAS10840
Anti-IgG (H+L) Donkey Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Mouse, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR