Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 594)

Dodavatel: Agrisera
AGRIAS122100EA 6150 CZK
AGRIAS122100
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 594)
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Donkey
Isotype:
Reactivity: Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR