Tisk…

Anti-Biotin Goat Polyclonal Antibody (DyLight® 650)

Dodavatel: Agrisera
AGRIAS122228EA 6940 CZK
AGRIAS122228
Anti-Biotin Goat Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Secondary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR