Tisk…
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 650)
Dodavatel: Agrisera
AGRIAS122253EA 6150 CZK
AGRIAS122253
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Mouse, Rabbit, Sheep