Tisk…
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 650)
Dodavatel: Agrisera
AGRIAS122298EA 7730 CZK
AGRIAS122298
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit