Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 650)

Dodavatel: Agrisera
AGRIAS122329EA 6570 CZK
AGRIAS122329
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR