Tisk…

Anti-IL17A Mouse Monoclonal Antibody (Unconjugated) [clone: 4H1524.1]

Dodavatel: Rockland Immunochemicals
ROCK209-301-B32SEA 3010 CZK
ROCK209-301-B32S
Anti-IL17A Mouse Monoclonal Antibody (Unconjugated) [clone: 4H1524.1]
Protilátky
Il-17A antibody has been tested for use in ELISA and western blotting. By western blot a band approximately ~17.5 kDa in size corresponding to human IL17-A Protein is expected in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 4H1524.1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR