Tisk…

Anti-AKT1 Mouse Polyclonal Antibody (Unconjugated) [clone: 17F6.B11]

Dodavatel: Rockland Immunochemicals
ROCK200-301-268EA 11200 CZK
ROCK200-301-268
Anti-AKT1 Mouse Polyclonal Antibody (Unconjugated) [clone: 17F6.B11]
Protilátky
Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract. This phospho-specific monoclonal antibody reacts with human and mouse AKT pS473.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500-1:3000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: 17F6.B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR