Tisk…

Anti-MAD1L1 Mouse Polyclonal Antibody (Unconjugated) [clone: 9B10]

Dodavatel: Rockland Immunochemicals
ROCK200-301-903EA 22480 CZK
ROCK200-301-903
Anti-MAD1L1 Mouse Polyclonal Antibody (Unconjugated) [clone: 9B10]
Protilátky
This Protein A purified antibody has been tested for use in flow cytometry, immunoprecipitation, immunofluorescence and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band at ~ 83 kDa.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MAD1L1
Clonality: Polyclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR