Tisk…

Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 39A1413]

Dodavatel: Rockland Immunochemicals
ROCK200-303-382EA 21190 CZK
ROCK200-303-382
Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 39A1413]
Protilátky
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 39A1413
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR