Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Dodavatel: Rockland Immunochemicals
ROCK714-408-002EA 13150 CZK
ROCK714-408-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Protilátky
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 500 µl
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR