CF™488A antibodies are affinity-purified antibodies labeled with green fluorescent CF™488A, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™488A is a green fluorescent dye with excitation/emission maxima at 490/515nm. CF™488A is spectrally similar to AlexaFluor®488, ATTO 488, Cy®2, DyLight®488, and fluorescein (FAM or FITC). CF™488A is exceptionally bright and its fluorescence is insensitive to pH. Moreover, CF™488A is far more photostable than FITC or fluorescein. The slightly shorter emission wavelength of CF™488A compared to other green dyes offers the advantage of less fluorescence spill-over in the red channel in multi-color detection applications.
CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. Cy® is a registered trademark of GE Healthcare. DyLight® is a registered trademark of Pierce Biotechnology.
Informace pro objednávání: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.
Type: Secondary
Antigen: IgM MU CHAIN
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human