Tisk…

Anti-MAD2L1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP20713CEA 9810 CZK
ABGEAP20713C
Anti-MAD2L1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR