Tisk…

Anti-SCUBE3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP19055CEA 9810 CZK
ABGEAP19055C
Anti-SCUBE3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SCUBE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR