Tisk…

Anti-MMD2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP18167AEA 9810 CZK
ABGEAP18167A
Anti-MMD2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MMD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR