Tisk…

Anti-CCDC155 Rabbit Polyclonal Antibody

Dodavatel: Abgent
ABGEAP20386CEA 9870 CZK
ABGEAP20386C
Anti-CCDC155 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CCDC155
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR