Tisk…

Anti-TAB3 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB22722EA 14770 CZK
ABNOPAB22722
Anti-TAB3 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant MAP3K7IP3.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: TAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR