Tisk…

Anti-CDCA8 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB15529EA 18330 CZK
ABNOPAB15529
Anti-CDCA8 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against synthetic peptide of CDCA8.

Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Western Blot: 2 ?g/ml

Type: Primary
Antigen: CDCA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR