Tisk…

Anti-SRP72 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB22716EA 14770 CZK
ABNOPAB22716
Anti-SRP72 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant SRP72.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: SRP72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR