Tisk…

Anti-PURH Mouse Monoclonal Antibody [clone: F38P7H9]

Dodavatel: Abnova
ABNOMAB7973EA 21890 CZK
ABNOMAB7973
Anti-PURH Mouse Monoclonal Antibody [clone: F38P7H9]
Mouse monoclonal antibody raised against synthetic peptide of ATIC.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 15 ?g/ml

Type: Primary
Antigen: PURH
Clonality: Monoclonal
Clone: F38P7H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Frog, Drosophila, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR