|
|
OBJEDNÁVKA
Produktová specifikace
Symbol antigenu | CCT2 |
Název antigenu | Chaperonin Containing TCP1, subunit 2 (beta) |
Reaktivita | Frog, Rat, Mouse, Human |
Spojení | Unconjugated |
Klonovalita | Monoclonal |
Druh protilátky | Primary |
Klon | F39P7F11 |
Hostitel | Mouse |
Isotyp | IgG2a |
Western blot | Yes |
ImmunoChemistry | Yes |
Synonyma antigenu | Chaperonin containing TCP1 subunit 2, CCT-beta, TCPB_HUMAN, Chaperonin containing t complex polypeptide 1 subunit 2, CTP:phosphocholine cytidylyltransferase 2, T complex protein 1 subunit beta, CCT-2, MGC142074, T complex protein 1 beta subunit, MGC94480, T-complex protein 1 subunit beta, Chaperonin containing TCP1 subunit 2 beta, TCP 1 beta, CCT beta, PRO1633, MGC142076, Chaperonin containing t complex polypeptide 1 beta subunit, 99D8.1., TCP-1-beta, CCTB, CCT 2 |
Gene ID | 10576 |
Formát | Liquid |
Křížová adsorbce | No |
Imunogen | A synthetic peptide (conjugated with ovalbumin) corresponding to human CCT2. |
Skladovací pufr | phosphate-buffered solution, pH 7,2 (0,09% sodium azide) |
Skladovací teplota | Store at 4 °C for three months. For long term storage store at – 20 °C. Aliquot to avoid repeated freezing and thawing. |