Tisk…

Anti-GSE1 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB22710EA 14770 CZK
ABNOPAB22710
Anti-GSE1 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant KIAA0182.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: GSE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR