Tisk…

Anti-PID1 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB22705EA 14770 CZK
ABNOPAB22705
Anti-PID1 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant PID1.

Recommended Dilutions: Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR