Tisk…

Anti-PDK4 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB15513EA 18330 CZK
ABNOPAB15513
Anti-PDK4 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against synthetic peptide of PDK4.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: PDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR