Tisk…

Anti-SCD Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB15510EA 18330 CZK
ABNOPAB15510
Anti-SCD Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against synthetic peptide of SCD.

Recommended Dilutions: Immunocytochemistry: 1:50; Immunofluorescence: 1:50; Western Blot: 2 ?g/ml

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR