Tisk…

Anti-TCF7L2 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB15508EA 18330 CZK
ABNOPAB15508
Anti-TCF7L2 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against synthetic peptide of TCF7L2.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: TCF7L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR