Tisk…

Anti-ALS2CR12 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB22697EA 14770 CZK
ABNOPAB22697
Anti-ALS2CR12 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against recombinant ALS2CR12.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: ALS2CR12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR