Tisk…

Anti-Hepatitis C virus NS5b Protein Mouse Monoclonal Antibody [clone: NS5B-6]

Dodavatel: Abnova
ABNOMAB7950EA 10150 CZK
ABNOMAB7950
Anti-Hepatitis C virus NS5b Protein Mouse Monoclonal Antibody [clone: NS5B-6]
Protilátky
Mouse monoclonal antibody raised against recombinant hepatitis C virus NS5b protein.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: NS5B-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Virus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR