Tisk…

Anti-Cnga1/Cnga3 Mouse Monoclonal Antibody (ATTO 488) [clone: S36-12]

Dodavatel: Abnova
ABNOMAB11694EA 15540 CZK
ABNOMAB11694
Anti-Cnga1/Cnga3 Mouse Monoclonal Antibody (ATTO 488) [clone: S36-12]
Protilátky
Mouse monoclonal antibody raised against partial synthetic protein of Cng.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Cnga1/Cnga3
Clonality: Monoclonal
Clone: S36-12
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR