Tisk…
Anti-CACNA1G Mouse Monoclonal Antibody [clone: S178A-9]
Dodavatel: Abnova
ABNOMAB11548EA 18330 CZK
ABNOMAB11548
Anti-CACNA1G Mouse Monoclonal Antibody [clone: S178A-9]
Protilátky
Mouse monoclonal antibody raised against recombinant of Cacna1g.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat