Tisk…
E-Z 96® Plant RNA kit
Dodavatel: VWR - Omega Bio-Tek
R1027-01EA 21220 CZK
OMEGR1027-01 OMEGR1027-02
E-Z 96® Plant RNA kit
Based on the same principle of E.Z.N.A.® Plant RNA Kit, the E-Z 96® Plant RNA kit is designed for isolation of plant RNA in a 96-well plate format. The E-Z 96® Plant RNA Kit provides a convenient and rapid method for the isolation of total RNA from a variety of plant samples. The kit includes a homogenization plate to efficiently remove cell debris and simultaneously homogenize the lysate. In combination with the HiBind® RNA plate, this kit permits purification of high-quality RNA from as much as 40 mg seed tissue or up to 100 mg plant tissue. The system is also efficient enough to allow isolation of total RNA from as little as 0,5 mg tissue. Purified RNA is suitable for RT-PCR, Northern analysis, differential display, and poly A+ RNA selection.