Tisk…
Data logger for temperature and RH, HM9000
Dodavatel: Industrial Physics
SIMEHM9000EA 3280 CZK
SIMEHM9000
Data logger for temperature and RH, HM9000
This USB data logger is designed for the temperature and relative humidity measurements. Ideal for wide measurements in coating industry, food, hospital and construction/building maintanence. The measuring interval can be set to 2, 5, 10 or 30 seconds; 1, 5, 10 or 30 minutes; 1, 2, 3, 6, 12 or 24 hours.
  • Dew point and status indication
  • Long life battery with USB interface
  • Multimode to start logging
  • User-selectable alarm
  • Battery life depends on interval time settings
Informace o dodávce: Data logger is supplied with analysis software.