Tisk…
PCR master mix and hot start mix, peqGOLD
PEQL01-1499EA 0 CZK
PEQL01-1499 PEQL01-1699 PEQL01-1599
PCR master mix and hot start mix, peqGOLD
Prepared in 2X concentration, peqGOLD PCR Master Mixes contain an optimised mix of reaction buffer and nucleotides in addition to peqGOLD Taq DNA Polymerase. The Y mixes contain specifically optimised MgCl₂ concentrations for outstanding high yields. The S Mixes are optimised for high specificity PCR. All peqGOLD Master Mixes are designed for convenient handling, reduced risk of contamination and ensure the highest level of reproducibility.