Blood DNA Mini Kit Plus offers a safe, quick and efficient method for extracting pure and high quality DNA from fresh, frozen and anticoagulated treated blood.
- Extraction of DNA up to 60 kbp
- Suitable for human or animal blood
- Suitable for the simultaneous processing of multiple samples
- No need for time-consuming alcohol precipitations
- Complete enzymatic degradation of cellular RNA
- Efficient separation of enzyme inhibitors and contaminants
- Final elution of DNA in low salt buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids.
Informace o dodávce: Kits contain PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, Lysis Buffer BL, Binding Solution BL and TE Buffer, Proteinase K, DNA Wash Buffer BL I and II, and Elution Buffer.