Plant DNA Mini Kit for fast and efficient extraction of high quality DNA from nucleus, plastids and mitochondria of a wide range of plant species and plant tissues.
- Extraction of DNA up to 60 kbp
- Efficient separation of enzyme inhibitors and contaminants
- No need for organic extractions
- No need for time consuming alcohol precipitations
- Complete enzymatic degradation of the cellular RNA
- Final elution with low salt buffer or water
*Classic Line columns without lids.
Informace o dodávce: Kit comprises PerfectBind DNA columns (Classic Line), collection tubes, microfilter, Lysis Buffer PL1, Lysis Buffer PL2, DNA Binding Buffer, DNA Wash Buffer, and Elution Buffer.