Blood RNA Kits for quick isolation of high purity total RNA from fresh or anti-coagulated treated blood, including EDTA, citrate dextrose or heparin.
- DNA Removing columns for efficient separation of DNA and cellular debris
- No need for organic extractions or time-consuming alcohol precipitations
- Immediate inactivation of endogenous and exogenous RNases
- Efficient separation of enzyme inhibitors and contaminants
- Final elution with RNase-free water
*Classic Line columns without lids.
Informace o dodávce: Each kit contains PerfectBind RNA Safety Line columns, collection tubes, DNA Removing columns, Lysis Buffer ER, RNA Lysis Buffer T, RNA Wash Buffer I and II, and RNase-free water.