Tisk…
Blood RNA kits, peqGOLD
PEQLEMR511050EA 4450 CZK
PEQLEMR511050 PEQLEMR511200 PEQL12-6814-02 PEQLEMR511005 PEQL12-6814-01
Blood RNA kits, peqGOLD
Blood RNA Kits for quick isolation of high purity total RNA from fresh or anti-coagulated treated blood, including EDTA, citrate dextrose or heparin.
  • DNA Removing columns for efficient separation of DNA and cellular debris
  • No need for organic extractions or time-consuming alcohol precipitations
  • Immediate inactivation of endogenous and exogenous RNases
  • Efficient separation of enzyme inhibitors and contaminants
  • Final elution with RNase-free water
*Classic Line columns without lids.
Informace o dodávce: Each kit contains PerfectBind RNA Safety Line columns, collection tubes, DNA Removing columns, Lysis Buffer ER, RNA Lysis Buffer T, RNA Wash Buffer I and II, and RNase-free water.