Tisk…
Total RNA Kit, peqGOLD
PEQL12-6634-01EMEA 4230 CZK
PEQL12-6634-01EM PEQL12-6634-00EM PEQLEMR505005 PEQLEMR505200 PEQL12-6634-00 PEQL12-6634-01 PEQL12-6634-02 PEQLEMR505050
Total RNA Kit, peqGOLD
Total RNA Kits for the quick and easy isolation of high quality total RNA from animal or human cells and tissue.
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions or time-consuming alcohol precipitations
  • Immediate inactivation of exogenous and endogenous RNases
  • Favoured binding of RNA molecules >200 bases
  • Efficient separation of enzyme inhibitors and contaminants
  • Final elution in RNase-free water
  • Isolated total RNA directly suitable for downstream applications
*Safety Line columns with snap-on lids. Classic Line columns without lid.
Informace o dodávce: Each kit contains PerfectBind RNA columns (Safety or Classic Line*), collection tubes, DNA Removing columns, RNA Lysis Buffer T, RNA Wash Buffer I and II, and RNase-free water. Advanced protocol for isolation of miRNA included in the manual.