Tisk…
Anti-ZSCAN18 Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00065982-AP22EA 23800 CZK
ABNOH00065982-AP22
Anti-ZSCAN18 Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human ZSCAN18.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ZSCAN18 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-ZSCAN18 (20 µg);
Type:
Antigen: ZSCAN18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human