Tisk…

Anti-AHNAK Polyclonal Antibody Pair

Dodavatel: Abnova
ABNOH00079026-AP22EA 23090 CZK
ABNOH00079026-AP22
Anti-AHNAK Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human AHNAK.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AHNAK (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-AHNAK (20 µg);

Type:
Antigen: AHNAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR