Tisk…

Anti-PEPD Polyclonal Antibody Pair

Dodavatel: Abnova
ABNOH00005184-AP22EA 23090 CZK
ABNOH00005184-AP22
Anti-PEPD Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human PEPD.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-PEPD (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-PEPD (20 µg);

Type:
Antigen: PEPD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR