Tisk…
Anti-PDCD6IP Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00010015-PW1EA 23800 CZK
ABNOH00010015-PW1
Anti-PDCD6IP Polyclonal Antibody Pair
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-PDCD6IP (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-PDCD6IP (50 µl)
Type:
Antigen: PDCD6IP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human