Tisk…

Anti-ZNF263 Polyclonal Antibody Pair

Dodavatel: Abnova
ABNOH00010127-AP22EA 23800 CZK
ABNOH00010127-AP22
Anti-ZNF263 Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human ZNF263.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ZNF263 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-ZNF263 (20 µg);

Type:
Antigen: ZNF263
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR