Tisk…
Anti-HKDC1 Mouse Monoclonal Antibody
Dodavatel: Abnova
ABNOH00080201-PW1EA 23800 CZK
ABNOH00080201-PW1
Anti-HKDC1 Mouse Monoclonal Antibody
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-HKDC1 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-HKDC1 (50 ?g)
Type:
Antigen: HKDC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human