Tisk…
Anti-SCLY Mouse Monoclonal Antibody
Dodavatel: Abnova
ABNOH00051540-PW2EA 23090 CZK
ABNOH00051540-PW2
Anti-SCLY Mouse Monoclonal Antibody
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-SCLY (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-SCLY (50 ?g)
Type:
Antigen: SCLY
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human