Tisk…

Anti-GTF3C5 Mouse Monoclonal Antibody

Dodavatel: Abnova
ABNOH00009328-PW1EA 23090 CZK
ABNOH00009328-PW1
Anti-GTF3C5 Mouse Monoclonal Antibody
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-GTF3C5 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-GTF3C5 (50 ?g)

Type:
Antigen: GTF3C5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR