Tisk…

Anti-BLNK Antibody Pair

Dodavatel: Abnova
ABNOH00029760-AP51EA 23090 CZK
ABNOH00029760-AP51
Anti-BLNK Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human BLNK.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-BLNK, IgG2a kappa (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-BLNK (50 µg);

Type:
Antigen: BLNK
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR