Tisk…
Anti-FGL2 Polyclonal Antibody Pair
Dodavatel: Abnova
ABNOH00010875-PW2EA 23800 CZK
ABNOH00010875-PW2
Anti-FGL2 Polyclonal Antibody Pair
Protilátky
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-FGL2 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-FGL2 (50 µg)
Type:
Antigen: FGL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human