Tisk…

Anti-IL1B Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Abnova
ABNOH00003553-AP31EA 23090 CZK
ABNOH00003553-AP31
Anti-IL1B Rabbit Polyclonal Antibody (Biotin)
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human IL1B.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-IL1B (100 ?g); 2. Detection Antibody: biotinylated Rabbit MaxPab® affinity purified Polyclonal Anti-IL1B (20 ?g);

Type:
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR