Tisk…

Anti-IL1B Polyclonal Antibody Pair

Dodavatel: Abnova
ABNOH00003553-AP91EA 23090 CZK
ABNOH00003553-AP91
Anti-IL1B Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human IL1B.

Antibody pair set content: 1. Capture Antibody: Mouse purified Polyclonal Anti-IL1B (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-IL1B (50 µg);

Type:
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR