Tisk…

Anti-CD40L Antibody Pair

Dodavatel: Abnova
ABNOH00000959-AP13EA 23090 CZK
ABNOH00000959-AP13
Anti-CD40L Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human CD40LG.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CD40LG (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-CD40LG, IgG2b kappa (20 µg);

Type:
Antigen: CD40L
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR